• USA Home
  • Z351881 - Sigma-Aldrich® nucleic acid sequencing units
EMAIL THIS PAGE TO A FRIEND
Z351881 Sigma
Sigma-Aldrich® nucleic acid sequencing units
Adjustable Height, Dual Gel, gel W × H 20 cm × 22-62 cm
Purchase
Properties
Related Categories Electrophoresis Supplies, Labware, Units /Gel Box, Vertical Gel Electrophoresis Units
availability   not available in EU
gel W × H   20 cm × 22-62 cm
Description
Features and Benefits
Designed to run 2gels at the same time, any length from 22 to 62cm. The upper buffer reservoir slides on two 1-inch Delrin rods, secures by stainless steel thumbscrews, and serves both sides; the bottom reservoirs are independent. Upper and lower reservoirs are removable. Features a convenient rotating base. Supplied with 2sets of 62cm glass plates. Cooling plates not included.
Special features of these units are:
• safety interlock lids
• detachable upper and lower buffer reservoirs
• upper and lower reservoirs have drains with tubing adapters for easy and safe disposal of buffer solutions
• leveling base with bubble level
Each unit comes complete with clamps, sealing tape, power leads, and glass plates.
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?