• USA Home
  • Z352187 - Combs for Sigma-Aldrich® sequencing units
EMAIL THIS PAGE TO A FRIEND
Z352187 Sigma
Combs for Sigma-Aldrich® sequencing units
size 0.2 mm × 94 sharktooth wells, for 33 cm wide gel
Purchase
Properties
Related Categories Combs for Sigma-Aldrich sequencing units, Electrophoresis Supplies, Labware, Sigma-Aldrich Vertical Electrophoresis Combs, Vertical Gel Electrophoresis Units More...
availability   not available in EU
size   0.2 mm × 94 sharktooth wells, for 33 cm wide gel
Description
Application
Precision-machined polycarbonate combs for sample well formation.
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?