• USA Home
  • Z352659 - Aluminum cooling plates for Sigma-Aldrich® sequencing units
EMAIL THIS PAGE TO A FRIEND
Z352659 Sigma
Aluminum cooling plates for Sigma-Aldrich® sequencing units
For 20 × 32 cm gels
Purchase
Description
Features and Benefits
Made from 1/4 in. aluminum with electrically insulating coating. Plate provides full-width cooling for 20cm or 33cm wide gels. Plate is approximately 16cm shorter than the glass plate to leave room for the upper buffer reservoir. Needed only for adjustable height sequencing units.
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?