• USA Home
  • Z352802 - Vertical electrophoresis unit, adjustable height, dual gel
EMAIL THIS PAGE TO A FRIEND
Z352802 Sigma
Vertical electrophoresis unit, adjustable height, dual gel
gel W × H 16.5 cm × 14.5-28 cm, Not CE-approved
Purchase
Properties
Related Categories Electrophoresis Supplies, Labware, Sigma-Aldrich Vertical Electrophoresis Units, Units /Gel Box, Vertical Gel Electrophoresis Units More...
availability   not available in EU
gel W × H   16.5 cm × 14.5-28 cm
Description
Features and Benefits
• Made from high quality acrylic with platinum electrodes
• Long lasting silicone sponge gasket on upper reservoir for leak-free operation without grease
• Safety covers with attached safety-tip power leads
• Designed for use with unique gel-casting gasket; heavy-duty casting clamps compress gasket between glass plates, sealing without grease
For Slab Gel Electrophoresis:
• Designed to use as little as 200 mL of buffer, and as little as 5 μg of sample per well
• Use with gel casting accessories which offer the ability to cast one or two gels in a single assembly
• Use in standard slab gel electrophoresis or in the second dimension of 2D electrophoresis
Each unit comes with base unit, upper and lower reservoirs with safety covers and attached power leads, and four gel-mounting spring clamps.
CE-APPROVED ONLY WHERE INDICATED
Slab and tube gel accessories are not included; order from the accessories list
General description
Not CE approved
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?