• USA Home
  • Z352829 - Multiple mini-gel casting chambers
EMAIL THIS PAGE TO A FRIEND
Z352829 Sigma
Multiple mini-gel casting chambers
Purchase
Description
General description
Cast multiple gels simultaneously. Central filling trough in bottom of chamber connects to a back fill port and assures even casting for single percentage or gradient gels. A silicone gasket, stainless steel spring clamps, and Mylar® shim sheets are included.
Legal Information
Mylar is a registered trademark of Dupont Teijin Films
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?