EMAIL THIS PAGE TO A FRIEND
Z352837 Sigma
Wet Blotters
Standard Blotter
Purchase
Description
General description
Wet blotters are designed for Western, Southern, or Northern Blotting. Proteins and nucleic acids from agarose or acrylamide gels can be transferred onto nitrocellulose, nylon, DEAE, or other derivatized paper.
The Mini and Standard blotters each include removable platinum electrodes, a safety cover, and power leads. The standard blotter includes one gel cassette, while the mini blotter includes four gel cassettes. The cassettes include two Scotch-Brite pads and two hinged, perforated fiberglass panels.
The Standard-blotter can hold up to three cassettes and the Mini-blotter can hold up to four cassettes allowing for multiple transfers at one time. The Mini-blotter features an internal glass cooling base. The cooling tube for the Standard-blotter must be ordered separately.
The Mini-blotter cassette can accommodate gels up to 10 × 10 cm. The Standard-blotter can accommodate gels up to 15 × 21.5 cm.
The Mini wet blotter is designed for Western blots.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
EP1503 Sigma-Aldrich® SB10 blotting system mini cassette, 10 x 10cm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?