• USA Home
  • Z353019 - Millipore® Millicell® cell culture plate inserts
EMAIL THIS PAGE TO A FRIEND
Z353019 Sigma
Millipore® Millicell® cell culture plate inserts
pore size 0.4 μm, diam. 30 mm, non-treated
Purchase
Properties
mfr. no.   Millipore, PIHT03050
diam.   30 mm
pore size   0.4 μm
Description
General description
Membrane-based devices for use with standard plastic tissue culture plates. Inserts are made of a polycarbonate membrane in polystyrene plastic holders.
Tissue culture treated for attachment dependent cells or Non-Treated for suspension cells.
Legal Information
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?