• USA Home
  • Z353051 - Millipore® Millicell® cell culture plate inserts

EMAIL THIS PAGE TO A FRIEND
Z353051 Sigma

Millipore® Millicell® cell culture plate inserts

pore size 3.0 μm, diam. 12 mm, tissue-culture treated

Purchase

Properties

Related Categories Cell Culture Supplies, Culture Plate Inserts, Labware, Manufacturer Browser, Millicell inserts,
material   polycarbonate filter
  polycarbonate plate
sterility   sterile
packaging   case of 50 ea for 24-well plate
mfr. no.   Millipore, PITP01250
diam.   12 mm
filter area   0.6 cm2
filter diam.   13 mm
size   24 wells
color   clear
matrix active group   Tissue culture treated surface treatment
pore size   3.0 μm

Description

General description

Membrane-based devices for use with standard plastic tissue culture plates. Inserts are made of a polycarbonate membrane in polystyrene plastic holders.
Tissue culture treated for attachment dependent cells or Non-Treated for suspension cells.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?