• USA Home
  • Z353086 - Millipore® Millicell® cell culture plate inserts

EMAIL THIS PAGE TO A FRIEND
Z353086 Sigma

Millipore® Millicell® cell culture plate inserts

pore size 0.4 μm, diam. 30 mm, tissue-culture treated

Purchase

Properties

Related Categories Cell Culture Supplies, Culture Plate Inserts, Labware, Manufacturer Browser, Millicell inserts,
material   polycarbonate plate
sterility   sterile
mfr. no.   Millipore, PIHP03050
diam.   30 mm
size   6 wells
color   clear
matrix active group   Tissue culture treated surface treatment
pore size   0.4 μm

Description

General description

Membrane-based devices for use with standard plastic tissue culture plates. Inserts are made of a polycarbonate membrane in polystyrene plastic holders.
Tissue culture treated for attachment dependent cells or Non-Treated for suspension cells.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?