• USA Home
  • Z353418 - Corning® 50 mL PP centrifuge tubes, self-standing
EMAIL THIS PAGE TO A FRIEND
Z353418 Sigma
Corning® 50 mL PP centrifuge tubes, self-standing
skirted with plug seal cap
Purchase
Properties
Related Categories 50 mL centrifuge tubes, Centrifugation Supplies, Centrifuge Tubes, Plastic, Corning, Corning Centrifuge Tubes,
material   cap (Plug Seal)
  polypropylene
  tube conical bottom
  clear tube
sterility   sterile
feature   RCF 3,000 × g
packaging   sleeve of 25
mfr. no.   Corning 430897
capacity   50 mL
Description
Legal Information
Corning is a registered trademark of Corning, Inc.
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?