• USA Home
  • Z353620 - Techne® hybridization tube
EMAIL THIS PAGE TO A FRIEND
Z353620 Sigma
Techne® hybridization tube
for Model HB-3D
Purchase
Properties
Related Categories Hybridization Ovens and Accessories, Hybridization Supplies, Labware, Molecular Biology Supplies
material   borosilicate glass
mfr. no.   Techne, FHB-16
Description
Compatibility
Push-on end cap seals suitable up to 70 °C only
Legal Information
Techne is a registered trademark of Techne, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?