• USA Home
  • Z353876 - SciLog Accu series Pumps
EMAIL THIS PAGE TO A FRIEND
Z353876 Sigma
SciLog Accu series Pumps
Model FM-40, AC input 220 V50 Hz
Purchase
Properties
feature   drive speed 0 ‑ 450 rpm
  flow range 0.2 ‑ 45 mL/min
  pressure 100 psi
mfr. no.   Scilog
availability   not available in EU
  not available in EU
Description
Features and Benefits
Pump head: valveless piston (RH1) for low-flow, high-precision metering; piston and chamber are ceramic, fittings and case are Kynar.
General description
Microprocessor controlled pump like the Expert series, but economical non-programmable alternative
• PumpSense safety feature prevents torque overload
• Corrosion-resistant housing
• Optically encoded, servo-controlled forward/reverse motor
• LED front panel displays pump speed
• Stackable up to three high
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?