• USA Home
  • Z354112 - Carestream® Kodak® autoradiography RP X-Omat developer/starter

EMAIL THIS PAGE TO A FRIEND
Z354112 Sigma

Carestream® Kodak® autoradiography RP X-Omat developer/starter

pack of 1 ea

Synonym: photo developing chemicals, photo processing chemicals

Purchase

Description

Compatibility

For automated processing systems

Quantity

Packet is sufficient to start 8 gal of developer.

Legal Information

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?