• USA Home
  • Z354120 - Carestream® Kodak® autoradiography developer system cleaner

EMAIL THIS PAGE TO A FRIEND
Z354120 Sigma

Carestream® Kodak® autoradiography developer system cleaner

pack of 1 gal

Synonym: photo developing chemicals, photo processing chemicals

Purchase

Description

Application

For cleaning developer racks and circulation systems.

Compatibility

For automated processing systems

Quantity

Liquid concentrate makes 6 gal.

Legal Information

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2922 8/PG 2
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?