• USA Home
  • Z354139 - Carestream® Kodak® autoradiography Fixer/Wash system cleaner

EMAIL THIS PAGE TO A FRIEND
Z354139 Sigma

Carestream® Kodak® autoradiography Fixer/Wash system cleaner

pack of 2.5 gal

Purchase

Description

Application

For cleaning fixer and wash racks, and fixer circulation systems.

Compatibility

For automated processing systems

Legal Information

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?