EMAIL THIS PAGE TO A FRIEND
Z354457 Sigma
Bar clamp
for 33 cm wide sequencing unit
Purchase
Description
Features and Benefits
Replaces bulldog spring clamps for attaching the gel sandwich to the upper reservoir. Gives even compression over entire width of gel, eliminating capillary leakage between samples and reducing glass plate fractures. Clamp is included with fixed-height units, but is optional with adjustable-height units.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?