• USA Home
  • Z354635 - Base Runner sequencing unit accessory
EMAIL THIS PAGE TO A FRIEND
Z354635 Sigma
Base Runner sequencing unit accessory
Comb/spacer set, size 0.4 mm
Purchase
Description
Features and Benefits
Includes four 32-rectangular-well combs, two 32-well and two 48-well sharktooth combs, eight side spacers, and four bottom spacers.
Legal Information
Base Runner is a trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?