• USA Home
  • Z354910 - Base Runner nucleic acid sequencing units
EMAIL THIS PAGE TO A FRIEND
Z354910 Sigma
Base Runner nucleic acid sequencing units
Base Runner 100 single-gel unit
Purchase
Description
Features and Benefits
These sequencers offer next-generation technology, with features that allow faster set-up and greater flexibility than conventional units. Summary of features:
• adjustable height and selection of accessories for running 30cm, 45cm, or 60cm gels; resolve more than 400 bases per run!
• patented glass-aluminum-glass laminated Thermocore™ backing plate efficiently distributes heat, reducing thermal artifacts.
• upper and lower buffer chambers are removable for convenient disposal of buffer; safety covers protect against electrical hazard
• spring clamp system and silicone gasket for clean, leak-free set-up
• recessed pure platinum electrodes for long life
General description
Unit includes one upper and one lower buffer reservoir, one Thermocore and one outer glass plate (60 cm), half of a 0.4 mm × 60 cm comb/spacer set (two 32-well conventional combs, one 32-well and one 48-well sharktooth combs, and four side spacers), and one set of power leads.
Legal Information
Base Runner is a trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?