• USA Home
  • Z354996 - Millicell® CM culture plate inserts

EMAIL THIS PAGE TO A FRIEND
Z354996 Sigma

Millicell® CM culture plate inserts

pore size 0.4 μm, diam. 30 mm

Purchase

Properties

Related Categories Cell Culture Supplies, Culture Plate Inserts, Labware, Manufacturer Browser, Millicell inserts,
material   polystyrene plate
sterility   sterile
mfr. no.   Millipore, PICM03050
diam.   30 mm
size   6 wells
color   clear
matrix active group   Non-treated surface treatment
pore size   0.4 μm

Description

General description

Transparent Biopore CM hydrophilic PTFE membrane in a polystyrene housing

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?