• USA Home
  • Z355097 - Millipore® Millicup filter units
EMAIL THIS PAGE TO A FRIEND
Z355097 Sigma
Millipore® Millicup filter units
Millicup HV
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millicup sterile filter units, Millipore,
material   PVDF membrane
  high-density polyethylene housing
sterility   non-sterile
feature   Low-extractable Durapore
packaging   pack of 10
mfr. no.   Millipore, SJHVM4710
funnel capacity   3 L
pore size   0.45 μm pore size
Description
General description
Disposable, polyethylene. Capacity is 300 mL, filter area 11.5 square cm.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?