• USA Home
  • Z355216 - Microcentrifuge Filters Ultrafree-DEAEMC
EMAIL THIS PAGE TO A FRIEND
Z355216 Sigma
Microcentrifuge Filters Ultrafree-DEAEMC
Purchase
Properties
feature   RCF 10,000 × g
mfr. no.   Millipore, UFC3DEA25
capacity   400 μL
storage temp.   2-8°C
Description
Features and Benefits
Protein capacity is >0.2 mg human serum albumin; pH range is 2-12.
Other Notes
DEAE-derivatized 1.2 μm cellulose acetate membrane mounted in an Ultrafree device.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?