• USA Home
  • Z355267 - Millipore® Sterivex filter unit
EMAIL THIS PAGE TO A FRIEND
Z355267 Sigma
Millipore® Sterivex filter unit
GV without filling bell, pore size 0.22 μm, Durapore PVDF membrane
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millipore, Millipore Filtration,
material   Durapore PVDF membrane
sterility   sterile
packaging   pack of 15
mfr. no.   Millipore, SVGV01015
pore size   0.22 μm
Description
General description
Suitable for sterilizing and clarifying tissue culture media and other aqueous solutions in the 100 mL to 1 liter volume range. Pressure-driven, self-venting, direct-dispensing design allows filtration into any type of container without foaming. Can be used with a peristaltic pump, pressure vessel, or syringe.
Connections are female luer-lock inlet and 3.3 mm O.D. male tubing outlet or male luer-lock outlet.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Sterivex is a trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?