• USA Home
  • Z355321 - Millipore® Sterivex filter unit
EMAIL THIS PAGE TO A FRIEND
Z355321 Sigma
Millipore® Sterivex filter unit
GS without filling bell, pore size 0.22 μm, mixed cellulose esters (HA) membrane
Purchase
Properties
material   mixed cellulose esters (HA) membrane
sterility   sterile
mfr. no.   Millipore, SVGS01015
pore size   0.22 μm
Description
General description
Suitable for sterilizing and clarifying tissue culture media and other aqueous solutions in the 100 mL to 1 liter volume range. Pressure-driven, self-venting, direct-dispensing design allows filtration into any type of container without foaming. Can be used with a peristaltic pump, pressure vessel, or syringe.
Connections are female luer-lock inlet and 3.3 mm O.D. male tubing outlet or male luer-lock outlet.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Sterivex is a trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z359912 Millipore® Sterivex filter unit, GP without filling bell, pore size 0.22 μm, polyethersulfone membrane
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?