• USA Home
  • Z355461 - Millipore® Sterifil® filtration system
EMAIL THIS PAGE TO A FRIEND
Z355461 Sigma
Millipore® Sterifil® filtration system
Complete Holder
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millipore, Millipore Filtration,
material   polypropylene housing
  polysulfone housing
sterility   sterile
feature   cap plug seal
packaging   pack of 1
mfr. no.   Millipore, XX1104710
Description
General description
A reusable, aseptic filter system that can be autoclaved with the filter of choice in place. Construction consists of a polysulfone funnel, funnel cover, and cover; polypropylene base and filter support screen; silicone stopper and O-ring. Accepts a 47 mm filter. Capacity is 250 mL.
Perforated stopper on holder outlet mounts in a standard 1-liter filtering flask or Millipore filtering manifold. Two ports on receiver flask accept either 6 mm (1/4 in.) I.D. tubing or male luer-slip connection for vacuum and drain or vent.
Four female luer-slip ports on funnel cover can be used for vent and aseptic inlet.
Includeds polysulfone funnel, funnel cover, receiver flask and cover; polypropylene holder base and filter support screen; silicone stopper
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Sterifil is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?