• USA Home
  • Z355488 - Millex® syringe filter units, disposable, Durapore® PVDF
EMAIL THIS PAGE TO A FRIEND
Z355488 Sigma
Millex® syringe filter units, disposable, Durapore® PVDF
pore size 0.1 μm, diam. 33 mm, γ-irradiated
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millex syringe filter units, disposable, Durapore PVDF, Millipore,
material   PVDF membrane
  modified acrylic housing
sterility   γ-irradiated
packaging   pack of 50 ea
mfr. no.   Millipore SLVV033RS
  Millipore, SLVV033RS
  Millipore, SLVVR25LS
diam.   33 mm
volume   ≤100 mL
pore size   0.1 μm
fitting   inlet female Luer (lock)
  outlet male Luer
Description
Application
Tissue culture; mycoplasma reduction
Ultra-low protein binding (<1 μg/cm2). Component materials meet USP XXI Class VI Biological Tests for plastics. Water extractables <0.5%
General description
Units contain Durapore Hydrophilic PVDF filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings.
Units smaller than 25 mm diameter are specifically designed to minimize hold-up in filtration of small volumes. The 3-4 mm units are recommended for volumes of <1 mL; the 13 mm units are recommended for volumes of 0.5 - 10 mL.
Legal Information
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
Millex is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Articles
Sterile Millex® Filters for Cell Culture
Millex syringe filters are ideal for sterilizing 1 mL – 200 mL of cell culture reagents, such as cell culture media, antibiotics, supplements, sera, viruses for infection, and other biological soluti...
Keywords: Antibiotics, Capillary electrophoresis, Cell culture, Culture media, Filtration, Growth factors, Pharmaceutical, Sterilizations
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?