Z355518 Sigma

Millex® syringe filter units, disposable, Durapore® PVDF

pore size 0.45 μm, filter diam. 33 mm, γ-irradiated

DOWNLOAD MSDS (PDF)

Properties

Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millex syringe filter units, disposable, Durapore PVDF, Millipore,
material   PVDF membrane
  modified acrylic housing
sterility   γ-irradiated
mfr. no.   Millipore SLHV033RS
  Millipore, SLHV033RS
  Millipore, SLHVR25LS
packaging   pack of 50 ea
filter diam.   33 mm
volume   ≤100 mL
pore size   0.45 μm
fitting   inlet female Luer (lock)
  outlet male Luer

Description

Application

Tissue culture media, serum, other additives; DNA; viruses

Ultra-low protein binding (<1 μg/cm2). Component materials meet USP XXI Class VI Biological Tests for plastics. Water extractables <0.5%

General description

Units contain Durapore Hydrophilic PVDF filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings. Units smaller than 25 mm diameter are specifically designed to minimize hold-up in filtration of small volumes. The 3-4 mm units are recommended for volumes of <1 mL; the 13 mm units are recommended for volumes of 0.5 - 10 mL.

Suitability

Suitable for filtration of biological samples.

Legal Information

Durapore is a registered trademark of Merck KGaA

Millex is a registered trademark of Merck KGaA

Price and Availability

Customers Also Viewed

Millex<SUP>®</SUP> syringe filter units, disposable, Durapore<SUP>®</SUP> PVDF

pore size 0.22 μm, diam. 33 mm, γ-irradiated

Millex<SUP>®</SUP> syringe filter units, disposable, Durapore<SUP>®</SUP> PVDF

pore size 0.45 μm, diam. 33 mm, non-sterile

Millex<SUP>®</SUP> syringe filter units, disposable, mixed esters

pore size 0.45 μm, diam. 33 mm

Millex<SUP>®</SUP> syringe filter units, disposable, PTFE

pore size 0.45 μm, diam. 13 mm, Millipore SLCR013NL, modified hydrophillic PTFE membrane

Millex<SUP>®</SUP> syringe filter units, disposable, PTFE

pore size 0.45 μm, diam. 25 mm, Millipore SLCR025NS, modified hydrophillic PTFE membrane

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?