• USA Home
  • Z355550 - Filter membranes for microbiological analysis
EMAIL THIS PAGE TO A FRIEND
Z355550 Sigma
Filter membranes for microbiological analysis
pore size 0.7 μm (HC), S-Pak, White with black grid surface
Purchase
Properties
Related Categories Filter membranes for microbiological analysis, Filtration, Filtration Supplies, Labware, Manufacturer Browser,
material   cellulose acetate/cellulose nitrate mixed esters both filters
mfr. no.   Millipore HCWG047S1
  Millipore, HCWG047S1
  Millipore, HCWG047S1
pore size   0.7 μm (HC)
Description
General description
Filter type HA, 0.45 μm pore size, is designed to give complete retention and maximum recovery of total coliforms and fecal coliform bacteria. Complies with applicable U.S. EPA Standard methods and ASTM specifications for membrane filters used for drinking water analysis.
Filter type HC, 0.7 μm pore size, is designed for improved recovery of stressed fecal coliform organisms, especially those found in chlorinated effluents. The larger pore size permits faster filtration if the water sample has a higher particulate burden.
Both filters are cellulose acetate/cellulose nitrate mixed esters.
S-Kit includes filter membrane plus absorbent pad, sterile, individually sealed, packed in 100′s in dispenser tubes.
S-Pak includes filter membrane, sterile, individually sealed.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?