• USA Home
  • Z356042 - Horizontal IBI electrophoresis units
EMAIL THIS PAGE TO A FRIEND
Z356042 Sigma
Horizontal IBI electrophoresis units
Model MP 1015, Multipurpose
Purchase
Description
Features and Benefits
includes two 2mm × 16-tooth combs
General description
These units are complete designs that incorporate the best features of the popular IBI units.
Features:
• Durable acrylic construction with fewer seams and more unitized parts
• Vented safety lid allows heat to escape and improves visibility by reducing condensation
• External casting system allows one gel to be cast (without tape) while another is running
• Electrical contact is made by gently placing the lid on the chamber, eliminating sample disturbance during set-up
• Gel trays are transparent to 312 nm UV light to allow visualization on the tray
• Dark stripes below each comb position make it easy to see wells in submerged gel
• Platinum electrodes and gold-plated contacts resist corrosion
Packaging
Each of these units comes complete and includes a casting system with UV-transparent gel tray, leveling bubble, power leads, 2 combs and buffer recirculating ports. Does not include extra gel-casting accessories, combs, or replacement parts.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?