• USA Home
  • Z356050 - Gel casting tray for IBI horizontal unit
EMAIL THIS PAGE TO A FRIEND
Z356050 Sigma
Gel casting tray for IBI horizontal unit
size 10 cm × 15 cm
Purchase
Description
Features and Benefits
Transparent to 312 nm UV, so gel does not have to be removed from tray for viewing. One included with each base unit.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?