• USA Home
  • Z356263 - Horizontal IBI electrophoresis units

EMAIL THIS PAGE TO A FRIEND
Z356263 Sigma

Horizontal IBI electrophoresis units

Model HR 2025, High Resolution

Purchase

Properties

Related Categories Electrophoresis Supplies, Horizontal Gel Electrophoresis Supplies, Horizontal IBI electrophoresis units, IBI, IBI Scientific,
feature   capacity 160 samples four 40-tooth combs
  comb positions 5
mfr. no.   IBI, IB56000
buffer capacity   1,200 mL
gel W × L   20 cm × 25 cm

Description

Features and Benefits

includes two 2mm × 20-tooth combs

General description

These units are complete designs that incorporate the best features of the popular IBI units.

Features:
• Durable acrylic construction with fewer seams and more unitized parts
• Vented safety lid allows heat to escape and improves visibility by reducing condensation
• External casting system allows one gel to be cast (without tape) while another is running
• Electrical contact is made by gently placing the lid on the chamber, eliminating sample disturbance during set-up
• Gel trays are transparent to 312 nm UV light to allow visualization on the tray
• Dark stripes below each comb position make it easy to see wells in submerged gel
• Platinum electrodes and gold-plated contacts resist corrosion

Packaging

Each of these units comes complete and includes a casting system with UV-transparent gel tray, leveling bubble, power leads, 2 combs and buffer recirculating ports. Does not include extra gel-casting accessories, combs, or replacement parts.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?