• USA Home
  • Z356298 - Gel casting system for IBI horizontal units
EMAIL THIS PAGE TO A FRIEND
Z356298 Sigma
Gel casting system for IBI horizontal units
size 20 cm × 25 cm
Purchase
Description
Features and Benefits
Accessory holds casting tray and positions comb for leak-free gel casting without tape. One included with each base unit.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?