• USA Home
  • Z358347 - Septi-Clean hand cleaner

EMAIL THIS PAGE TO A FRIEND
Z358347 Sigma

Septi-Clean hand cleaner

one case = 24 × 4 fl. oz.

Purchase

Properties

Related Categories Cleaners, Hand Cleaners, Labware More...

Description

Application

An instant skin disinfectant. Evaporates quickly and does not leave any residual fragrance. Non-drying to skin; hypoallergenic.

Packaging

One case = 24 × 4 fl. oz.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
73.4 °F
Flash Point(C) 
23 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?