• USA Home
  • Z358436 - Sigma ondisk CD-ROM for Windows® 3.1/95 and Macintosh System 7/8
EMAIL THIS PAGE TO A FRIEND
Z358436 Sigma
Sigma ondisk CD-ROM for Windows® 3.1/95 and Macintosh System 7/8
Purchase
Description
Compatibility
SYSTEM REQUIREMENTS: Windows 386 or better with at least 4 MB available RAM; approx. 2 MB of hard disk space; Windows 3.1 or 95.
::FBMacintosh: ::FM68020 or better with at least 4 MB available RAM; approx. 2 MB of hard disk space; System 7 or higher. A Folio ::s6INFOBASE::S7, Copyright 1993 Folio Corporation
Features and Benefits
The entire text of the Sigma General Catalog on disk in fully searchable form. Search for any word, partial word, or phrase, and refine searches with logical operators (AND - OR - NOT) to restrict or expand a request. Results of searches can be saved to disk for printing or importing into word processing programs.
Examples:
• Search for products described as substrates for a particular enzyme.
• Search for peptides by amino acid sequence or partial sequence
The text of the product listing will include, where available, molecular formula, formula weight, CAS number, Merck Index monograph number, Color Index number, Enzyme Commission Number, and RTECS number; these can all be used as search terms. The CD-ROM is fully hypertext-linked for every cross-reference in the printed catalog.
Legal Information
Windows is a registered trademark of Microsoft Corporation
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?