• USA Home
  • Z358495 - Carestream® Kodak® BioMax® XAR Film

EMAIL THIS PAGE TO A FRIEND
Z358495 Sigma

Carestream® Kodak® BioMax® XAR Film

size 9.5 in. (24 cm) × 12 in. (30 cm), Kodak XAR-5 150 1451

Synonym: autoradiography film, xray film

Purchase

Properties

Related Categories Autoradiography, Autoradiography Film, Carestream Kodak, Carestream Kodak Autoradiography, Carestream Kodak Biomax XAR film,
mfr. no.   Kodak XAR-5 150 1451
size   9.5 in. (24 cm) × 12 in. (30 cm)

Description

General description

• Coated with emulsion on both sides of the clear base.
• Recommended for chemiluminescence.
• Very versatile for autoradiography with high or low energy isotopes.
• Sensitivity can be increased with intensifying screens when using low-energy isotopes.
• When handling film in a darkroom, Kodak LED Safelight Darkroom Illuminator is recommended.
• All the XAR films are the same, just packaged differently.

Bulk packed

Footnote

Kodak X-OMAT AR film was reformulated to become BioMax XAR film.

Legal Information

BioMax is a registered trademark of Carestream Health, Inc.

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?