• USA Home
  • Z358681 - Millipore® glass-fiber filters, AP series
EMAIL THIS PAGE TO A FRIEND
Z358681 Sigma
Millipore® glass-fiber filters, AP series
Type AP 25, circles size 29.3 cm
Purchase
Properties
material   borosilicate with acrylic binder
mfr. no.   Millipore, AP2529325
circles size   29.3 cm
Description
Application
Same retention as Type AP20 but thicker, much higher capacity.
General description
AP Series filters are primarily designed as prefilters to retain contaminants that would quickly plug membrane filters.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?