• USA Home
  • Z358800 - MilliBlot kit Complete
EMAIL THIS PAGE TO A FRIEND
Z358800 Sigma
MilliBlot kit Complete
Purchase
Description
Features and Benefits
A three-in-one system for dot blot, slot blot, and nucleic acid transfers. the kit consists of a vacuum plenum base with O-ring seal, two clamps with integral rubber feet, and three interchangeable sample modules:
• a porous plate unit for transfers from agarose mini-gels includes a filter support, a rubber gel gasket, and a buffer fence to hold the gel
• a MilliBlot-D for 96-well dot blots includes a dot duct plate which supports the membrane and an 8 × 12-dot sample manifold. Each well holds up to 500 μl of sample and blots to a 6 mm dot.
• a MilliBlot-S for 48-sample slot blots includes a slot duct plate and an 8 × 6-slot sample manifold. Slot can hold up to 600 μl and concentrates the sample into a 0.75 × 7.5 mm slot.
Legal Information
MilliBlot is a trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?