• USA Home
  • Z358827 - Kodak® X-Omat RA Duplicating Film
EMAIL THIS PAGE TO A FRIEND
Z358827 Sigma
Kodak® X-Omat RA Duplicating Film
size 35 cm × 43 cm
Purchase
Properties
Related Categories Autoradiography, Autoradiography Film, Electrophoresis Supplies, Labware
mfr. no.   Kodak 138 9923
size   35 cm × 43 cm
Description
General description
Single-emulsion film on clear base. Designed specifically for 1:1 duplication of autoradiograms or stained electrophoresis gels. Create contact prints with a radiographic duplicating unit or by direct exposure to UV light such as BLB fluorescent lamps. Can be processed with the same cycles and chemicals as original films.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?