• USA Home
  • Z359300 - SDS-Polyacrylamide Gel Electrophoresis: A Video Guide
EMAIL THIS PAGE TO A FRIEND
Z359300 Sigma
SDS-Polyacrylamide Gel Electrophoresis: A Video Guide
VHS/PAL
Purchase
Properties
publication info   M. Basta, Cold Spring Harbor Laboratory, Cold Spring Harbor, NY, 1994
mfr. no.   Cold Spring Harbor
Description
General description
This video provides a step-by-step guide to the successful creation of SDS-PAGE minigels. It covers: making stock solutions; assembling and disassembling apparati; casting, pouring, and running gels; and detecting proteins and determining molecular weights. Emphasis is placed on troubleshooting, with the inclusion of numerous technical tips that can make the difference between success and failure and hihgh and low-qulaity results. (34 minutes)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?