• USA Home
  • Z359327 - Hybridomas and Monoclonal Antibodies
EMAIL THIS PAGE TO A FRIEND
Z359327 Sigma
Hybridomas and Monoclonal Antibodies
VHS PAL
Purchase
Properties
Related Categories Books and Software, Immunology Books, Labware More...
publication info   C. Brinton, Cold Spring Harbor Laboratory, Cold Spring Harbor, NY, 1994
mfr. no.   Cold Spring Harbor
Description
General description
This video guide, produced at the University of Pittsburgh Hybridoma Center, demonstrates the technical steps required for the successful production of antibodies: immunization and bleeding of mice, fusion of spleen cells and myeloma cells with PEG and DMSO, HAT selection and screening off positive cultures and growth of hybridoma lines. The demonstrations are accompanied by audio commentary on the procedures involved and their theoretical basis. (32 minutes)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?