• USA Home
  • Z359475 - Probe-Clip press-seal incubation chambers
EMAIL THIS PAGE TO A FRIEND
Z359475 Sigma
Probe-Clip press-seal incubation chambers
L × W 40 mm × 22 mm, chamber volume 200 μL
Purchase
Properties
L × W   40 mm × 22 mm
chamber volume   200 μL
Description
General description
Probe-Clips are made of optically clear hydrophobic plastic bonded to a durable silicone gasket. A sealed chamber is formed when a Probe-Clip and a microscope slide are pressed together. Excellent for probe incubation, cytochemistry, blocking, etc. No evaporation, even for incubations of several days′ duration. Cross-contamination and spills are eliminated.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?