• USA Home
  • Z359599 - Targeted Mutagenesis in Mice: A Video Guide VHS/PAL
EMAIL THIS PAGE TO A FRIEND
Z359599 Sigma
Targeted Mutagenesis in Mice: A Video Guide VHS/PAL
Purchase
Properties
publication info   R.A. Pederson, et al., Cold Spring Harbor Laboratory Press, Cold Spring Harbor, NY, 1993
mfr. no.   Cold Spring Harbor
Description
Features and Benefits
Embryonic stem (ES) cells are important vehicles for transfer of genes into mice. This video provides step-by-step demonstrations of the following procedures required for making mutant mice: starting embryonic stem (ES) cell lines; making fibroblast feeder layers; growing established ES cell lines; targeting ES cell lines; making chimeras with ES cells; and making a mouse. (70 minutes)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?