• USA Home
  • Z359718 - Polaroid® GelCam® camera accessory
EMAIL THIS PAGE TO A FRIEND
Z359718 Sigma
Polaroid® GelCam® camera accessory
Electrophoresis hood with filter holder, diag. 7 in.
Purchase
Description
General description
Focal plane is recessed slightly inside the hood to compensate for typical gel thickness when hood rests on transilluminator.
Legal Information
GelCam is a registered trademark of Polaroid Corp.
Polaroid is a registered trademark of Polaroid Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?