• USA Home
  • Z359785 - Hanna HI 9321 pH meter
EMAIL THIS PAGE TO A FRIEND
Z359785 Sigma
Hanna HI 9321 pH meter
AC input 110 V
Purchase
Properties
feature   calibration automatic one- or two-point; preprogrammed with five standard buffers; READY indicator confirms calibration
  temperature compensation 0 ‑ 100 °C (manual or automatic with temperature probe included)
mfr. no.   Hanna
availability   not available in EU
measuring range   −1999-1999 mV ORP, resolution: 1 mV
  −399.9-399.9 mV ISE, resolution: 0.1 mV
  0-100 °C temperature, resolution: 0.1 °C
  0-14 pH, resolution: 0.01
Description
Features and Benefits
• Simultaneous display of pH/mV and °C
• Supplied complete with combination electrode (not recommended for use with Tris buffers), articulated electrode holder, dust cover, three buffers, and AC adapter
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?