• USA Home
  • Z360104 - Exposure cassette stainless steel with metal crossbar closure
EMAIL THIS PAGE TO A FRIEND
Z360104 Sigma
Exposure cassette stainless steel with metal crossbar closure
film W × L 7 in. (18 cm) × 17 in. (43 cm)
Purchase
Properties
Related Categories Autoradiography, Autoradiography Exposure Cassettes, Electrophoresis Supplies, Labware
material   metal cross-bar closure
  stainless steel
film W × L   7 in. (18 cm) × 17 in. (43 cm)
Description
General description
Rugged stainless steel holders for exposing sheet x-ray film during autoradiography. Riveted floating hinges and pressure from cross-bars maintain close contact between film and sample. Felt inner liner forms light-tight seal. Especially suited for use with intensifying screens or for exposures below 0 °C.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?