• USA Home
  • Z360228 - Nitrocellulose membranes, supported BA-S 85 0.45 μm pore size

EMAIL THIS PAGE TO A FRIEND
Z360228 Sigma

Nitrocellulose membranes, supported BA-S 85 0.45 μm pore size

size 15 cm × 15 cm

Purchase

Properties

Related Categories Hybridization Membranes, Hybridization Supplies, Labware, Molecular Biology Supplies
size   15 cm × 15 cm
suitability   suitable for Southern blots

Description

Application

These membranes have the same nucleic acid-binding capacity as standard nitrocellulose membranes and will easily stand repeated probing.

General description

Exceptionally resistant to tearing and cracking.
Membranes formed from nitrocellulose cast on a synthetic support

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?