• USA Home
  • Z360244 - Nalgene® magnetic culture vessel
EMAIL THIS PAGE TO A FRIEND
Z360244 Sigma
Nalgene® magnetic culture vessel
size 1 L
Synonym: hot plate sitrrer, magnetic stirrer, stirrer hot plate
Purchase
Properties
Related Categories Cell Culture Supplies, Labware, Manufacturer Browser, Nalgene, Nalgene Culture,
material   polycarbonate (autoclavable)
mfr. no.   Nalgene
size   1 L
Description
General description
Lightweight, break-resistant polycarbonate vessel specifically designed for culture on a magnetic stir plate. Suspended PTFE impeller gives efficient mixing at slow speed for less shear. Vessel bottom is shaped to prevent settling, and impeller height is adjustable. Two shoulder ports give access without interrupting incubation. Graduated to 1 L.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?