• USA Home
  • Z360309 - Safety wash bottles, Nalgene®
EMAIL THIS PAGE TO A FRIEND
Z360309 Sigma
Safety wash bottles, Nalgene®
capacity 250 mL
Purchase
Properties
Related Categories Bottles and Caps, Labware, Manufacturer Browser, Nalgene, Nalgene Bottles, Plastic,
material   low-density polyethylene bottle and tube
  polypropylene screw cap
description   Not recommended for use with acetone
mfr. no.   Nalgene
capacity   250 mL
Description
General description
One-piece bottle with molded-in delivery tube. No awkward delivery tube in cap to drip or become contaminated while bottle is being filled. Wide mouth for fast, safe filling. Guaranteed leakproof linerless closure. All red for easy identification and protection of contents from light. Useful with acids, bases, alcohols.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z329614 BRAND® wash bottles, LDPE, capacity 250 mL
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?