EMAIL THIS PAGE TO A FRIEND
Z360414 Sigma
Desiccator plate
diam. 140 mm, 8 holes
Purchase
Properties
mfr. no.   Nalgene
O.D.   230 mm
diam.   140 mm
Description
General description
This desiccator plate has a greater resistance to thermal shock than porcelain plates. It has a fire-polished surface, and is a light green ceramic bonded to a metal core for inertness and corrosion-resistance. Numbered quadrants are marked for easy location of crucibles, weighing bottles, and other containers. Recommended use with Nalgene desiccator such as D2547, Z354074 and D2797.
The center hole is 7/8 in. diam. The plates have 24 holes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?