• USA Home
  • Z361003 - Sigma adjustable volume multichannel pipette
EMAIL THIS PAGE TO A FRIEND
Z361003 Sigma
Sigma adjustable volume multichannel pipette
8 channel, capacity 5-50 μL
Purchase
Description
General description
Rugged, reliable pipettors for quick delivery into columns or rows of 96-well multiwell plates. Comfortable in the hand, with smooth plunger and convenient tip ejector. Rotating plunger sets volume.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?