• USA Home
  • Z361143 - Slide scanner, SprintScan 35
EMAIL THIS PAGE TO A FRIEND
Z361143 Sigma
Slide scanner, SprintScan 35
Conversion kit to Windows
Purchase
Properties
Related Categories Labware, Light Microscopy, Manufacturer Browser, Microscopy & Staining, Polaroid,
AC/DC input   110 / 220, 60-50
mfr. no.   Polaroid
availability   not available in EU
Description
Features and Benefits
Change platforms or provide dual-platform support for SprintScan. Kit includes SCSI cable, terminator, scanning software and interface card. See above for system requirements.
Create high-resolution digital scans of your 35mm slides in less than one minute. Just set desired resolution from 72 dpi − excellent for screen displays − to 2700 dpi − fine enough for full-page professional printing requirements. Then drop in the slide and point-and-click in the scanning utility software (included). SprintScan uses proprietary CCD technology to capture 24-bit RGB color in a single pass. Scans mounted slides and unmounted film or filmstrips (film holder included). Even negatives can be scanned and "reversed" to display as positive color.
Legal Information
SprintScan is a trademark of Polaroid Corp.
Price and Availability
Tissue Diagnostics
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?